3-Fluoro-5-(hydroxymethyl)benzeneboronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-5-(hydroxymethyl)phenylboronic acid

15575-1G 15575-25G 15575-500MG 15575-5G
76908-128EA 201.58 USD
76908-128 76908-130 76908-132 76908-134
3-Fluoro-5-(hydroxymethyl)benzeneboronic acid ≥95% research grade
3-Fluoro-5-(hydroxymethyl)benzeneboronic acid
Formula: C₇H₈BFO₃
MW: 169.946 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19687060
CAS Number: 1146614-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR