3,5-Dibromo-2-iodopyridine ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
15512-1G
15512-25G
15512-500MG
15512-5G
76908-104EA
260.99
USD
76908-104
76908-106
76908-108
76908-110
3,5-Dibromo-2-iodopyridine ≥97% research grade
3,5-Dibromo-2-iodopyridine
Formula:
C₅H₂Br₂IN MW: 362.79 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08543470 CAS Number: 436799-34-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...