3-Propoxy-2,4,6-trifluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15469-1G 15469-25G 15469-5G
76908-088EA 94.43 USD
76908-088 76908-090 76908-092
3-Propoxy-2,4,6-trifluorophenylboronic acid ≥97%
3-Propoxy-2,4,6-trifluorophenylboronic acid
Formula: C₉H₁₀BF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD07784389
CAS Number: 871125-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR