3-Benzyloxy-2-chloro-6-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Benzyoxy-2-chloro-6-fluorophenylboronic acid

15458-25G 15458-5G
76908-078EA 410.57 USD
76908-078 76908-080
3-Benzyloxy-2-chloro-6-fluorophenylboronic acid ≥97%
3-Benzyloxy-2-chloro-6-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BClFO₃
MW: 280.4900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878328
CAS Number: 957062-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR