3-Chloro-5-(dimethylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-5-(dimethylcarbonyl)phenylboronic acid, (3-Chloro-5-(dimethylcarbamoyl)phenyl)boronic acid

15433-1G 15433-25G 15433-500MG 15433-5G
76908-030EA 138.98 USD
76908-030 76908-032 76908-034 76908-036
3-Chloro-5-(dimethylcarbamoyl)phenylboronic acid ≥97%
3-Chloro-5-(dimethylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BClNO₃
MW: 227.4500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800861
CAS Number: 957120-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR