3-(Methylthio)-5-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15427-1G 15427-25G 15427-500MG 15427-5G
76908-014EA 274.78 USD
76908-014 76908-016 76908-018 76908-020
3-(Methylthio)-5-(trifluoromethyl)phenylboronic acid ≥97%
3-(Methylthio)-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD09800876
CAS Number: 957120-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR