4-(Difluoromethoxy)-3-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15414-1G 15414-25G 15414-500MG 15414-5G
76907-998EA 350.1 USD
76907-998 76908-000 76908-002 76908-004
4-(Difluoromethoxy)-3-fluorophenylboronic acid ≥97%
4-(Difluoromethoxy)-3-fluorophenylboronic acid
Formula: C₇H₆BF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD11036077
CAS Number: 958451-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR