3-Chlor-4-(isopropylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-4-(N-isopropylcarbamoyl)phenylboronic acid

15381-1G 15381-25G 15381-5G
76907-976EA 99.73 USD
76907-976 76907-978 76907-980
3-Chlor-4-(isopropylcarbamoyl)phenylboronic acid ≥97%
3-Chlor-4-(isopropylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD07363768
CAS Number: 1034102-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR