3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(3-Chloro-4-(cyclopropylcarbamoyl)phenyl)boronic acid, 3-Chloro-4-(cyclopropylcarbamoyl)benzeneboronic acid
15379-1G
15379-25G
15379-500MG
15379-5G
76907-964EA
162.32
USD
76907-964
76907-966
76907-968
76907-970
3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid ≥97%
3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₁BClNO₃ MW: 239.46 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07363770 CAS Number: 850589-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...