3-Chloro-4-(dimethylcarbamoyl)phenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3-Chloro-4-(N,N-dimethylcarbamoyl)phenylboronic acid, (3-Chloro-4-(dimethylcarbamoyl)phenyl)boronic acid, 3-Chloro-4-(dimethylcarbamoyl)benzeneboronic acid
15377-1G
15377-25G
15377-5G
76907-950EA
76.39
USD
76907-950
76907-952
76907-954
3-Chloro-4-(dimethylcarbamoyl)phenylboronic acid ≥97%
3-Chloro-4-(dimethylcarbamoyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₁BClNO₃ MW: 227.45 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07363772 CAS Number: 850589-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...