4-Bromo-2,5-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15345-1G 15345-25G 15345-5G
76907-888EA 84.88 USD
76907-888 76907-890 76907-892
4-Bromo-2,5-dimethylbenzaldehyde ≥97%
4-Bromo-2,5-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO
MW: 213.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16295236
CAS Number: 88111-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR