3-Bromo-2-chloro-5-fluorotoluene ≥97%

Supplier: AOBChem USA

Warning

15318-1G 15318-25G 15318-5G
76907-878EA 76.39 USD
76907-878 76907-880 76907-882
3-Bromo-2-chloro-5-fluorotoluene ≥97%
3-Bromo-2-chloro-5-fluorotoluene
Formula: C₇H₅BrClF
Storage Temperature: Refrigerator
MDL Number: MFCD09800774
CAS Number: 1000576-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR