3-Carboxy-5-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15313-25G 15313-5G
76907-866EA 375.56 USD
76907-866 76907-868
3-Carboxy-5-fluorophenylboronic acid ≥97%
3-Carboxy-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BFO₄
Storage Temperature: Ambient
MDL Number: MFCD07363791
CAS Number: 871329-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR