3-Bromo-5-chloro-2-fluorophenol ≥97% research grade

Supplier: AOBChem USA

Warning

15270-1G 15270-25G 15270-500MG 15270-5G
76907-830EA 281.14 USD
76907-830 76907-832 76907-834 76907-836
3-Bromo-5-chloro-2-fluorophenol ≥97% research grade
3-Bromo-5-chloro-2-fluorophenol
Formula: C₆H₃BrClFO
Storage Temperature: Ambient
MDL Number: MFCD28143932
CAS Number: 1804897-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR