4-Bromo-2-chloro-6-fluoroaniline ≥97%

Supplier: AOBChem USA

Warning

15223-25G 15223-5G
76907-810EA 244.01 USD
76907-810 76907-812
4-Bromo-2-chloro-6-fluoroaniline ≥97%
4-Bromo-2-chloro-6-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrClFN
MW: 224.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878163
CAS Number: 885453-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR