(3-Bromo-5-formyl-2-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-formyl-2-methoxyphenylboronic acid

15136-25G 15136-5G
76907-770EA 616.39 USD
76907-770 76907-772
(3-Bromo-5-formyl-2-methoxyphenyl)boronic acid ≥97%
(3-Bromo-5-formyl-2-methoxyphenyl)boronic acid
Formula: C₈H₈BBrO₄
MW: 258.86 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09265141
CAS Number: 1072951-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR