3-Chloro-(5-methoxycarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-5-(methoxycarbonyl)phenylboronic acid, (3-Chloro-5-(methoxycarbonyl)phenyl)boronic acid

15068-1G 15068-25G 15068-500MG 15068-5G
76907-732EA 192.03 USD
76907-732 76907-734 76907-736 76907-738
3-Chloro-(5-methoxycarbonyl)phenylboronic acid ≥97%
3-Chloro-(5-methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BClO₄
MW: 214.4107 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09475906
CAS Number: 957120-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR