3-Chloro-5-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

15053-25G 15053-5G
76907-712EA 325.7 USD
76907-712 76907-714
3-Chloro-5-(trifluoromethyl)phenol ≥97%
3-Chloro-5-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD04115852
CAS Number: 570391-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR