3-Cyano-4-fluorobenzoic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
14879-25G
14879-5G
76907-686EA
466.8
USD
76907-686
76907-688
3-Cyano-4-fluorobenzoic acid ≥95%
3-Cyano-4-fluorobenzoic acid
Formula:
C8H4FNO2 Storage Temperature: Ambient |
MDL Number:
MFCD03094339 CAS Number: 171050-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...