3-Bromo-5-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-(trifluoromethyl)benzeneboronic acid

14776-25G 14776-5G
76907-664EA 453.01 USD
76907-664 76907-666
3-Bromo-5-(trifluoromethyl)phenylboronic acid ≥97%
3-Bromo-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BBrF₃O₂
MW: 268.82
Storage Temperature: Ambient
MDL Number: MFCD08436051
CAS Number: 913835-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR