3,5-Difluoro-4-hydroxybenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

14616-25G 14616-5G
76907-646EA 268.41 USD
76907-646 76907-648
3,5-Difluoro-4-hydroxybenzoic acid ≥97% research grade
3,5-Difluoro-4-hydroxybenzoic acid
Formula: C₇H₄F₂O₃
MW: 174.102 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12406861
CAS Number: 74799-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR