3-Bromo-2-formylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

14584-1G 14584-25G 14584-500MG 14584-5G
76907-630EA 173.99 USD
76907-630 76907-632 76907-634 76907-636
3-Bromo-2-formylphenylboronic acid ≥95%
3-Bromo-2-formylphenylboronic acid
Formula: C₇H₆BBrO₃
Storage Temperature: Ambient
MDL Number: MFCD11856011
CAS Number: 928048-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR