3-Bromo-2-chloro-6-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

14539-1G 14539-25G 14539-500MG 14539-5G
76907-600EA 384.05 USD
76907-600 76907-602 76907-604 76907-606
3-Bromo-2-chloro-6-fluorophenol ≥97%
3-Bromo-2-chloro-6-fluorophenol
Formula: C₆H₃BrClFO
MW: 225.443 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20923580
CAS Number: 943830-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR