3-Bromo-5-fluoro-4-iodopyridine ≥97% research grade

Supplier: AOBChem USA

Warning

145294-1G 145294-500MG 145294-5G
76907-592EA 732.03 USD
76907-592 76907-594 76907-596
3-Bromo-5-fluoro-4-iodopyridine ≥97% research grade
3-Bromo-5-fluoro-4-iodopyridine
Formula: C₅H₂BrFIN
Storage Temperature: Refrigerator
CAS Number: 1349717-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR