3-Chloro-(4-ethoxycarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Chloro-4-(ethoxycarbonyl)phenyl)boronic acid, 3-Chloro-4-(ethoxycarbonyl)phenylboronic acid

14488-1G 14488-25G 14488-5G
76907-584EA 62.6 USD
76907-584 76907-586 76907-588
3-Chloro-(4-ethoxycarbonyl)phenylboronic acid ≥97%
3-Chloro-(4-ethoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BClO₄
MW: 228.4373 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659875
CAS Number: 850568-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR