3,6-Difluoro-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

14468-1G 14468-25G 14468-500MG 14468-5G
76907-574EA 231.28 USD
76907-574 76907-576 76907-578 76907-580
3,6-Difluoro-2-methoxybenzoic acid ≥97%
3,6-Difluoro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD08275290
CAS Number: 887267-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR