3-Chloro-4-hydroxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14459-25G 14459-5G
76907-568EA 183.54 USD
76907-568 76907-570
3-Chloro-4-hydroxyphenylboronic acid ≥97%
3-Chloro-4-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BClO₃
Storage Temperature: Ambient
MDL Number: MFCD09258739
CAS Number: 182344-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR