3-Cyano-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14441-25G 14441-5G
76907-560EA 410.57 USD
76907-560 76907-562
3-Cyano-2-fluorophenylboronic acid ≥97%
3-Cyano-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD07374984
CAS Number: 957121-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR