(3-Bromo-5-fluoro-2-isopropoxyphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-fluoro-2-isopropoxyphenylboronic acid

14414-25G 14414-5G
76907-540EA 418 USD
76907-540 76907-542
(3-Bromo-5-fluoro-2-isopropoxyphenyl)boronic acid ≥97% research grade
(3-Bromo-5-fluoro-2-isopropoxyphenyl)boronic acid
Formula: C₉H₁₁BBrFO₃
Storage Temperature: Ambient
MDL Number: MFCD06798051
CAS Number: 352534-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR