4-Bromo-2,6-dichlorobenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

14398-1G 14398-25G 14398-5G
76907-530EA 82.76 USD
76907-530 76907-532 76907-534
4-Bromo-2,6-dichlorobenzaldehyde ≥97% research grade
4-Bromo-2,6-dichlorobenzaldehyde
Formula: C₇H₃BrCl₂O
Storage Temperature: Refrigerator
MDL Number: MFCD11227147
CAS Number: 111829-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR