3-Methoxynaphthalen-2-ol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Hydroxy-2-methoxynaphthalene

14388-1G 14388-25G 14388-500MG 14388-5G
76907-518EA 201.58 USD
76907-518 76907-520 76907-522 76907-524
3-Methoxynaphthalen-2-ol ≥97%
3-Methoxynaphthalen-2-ol
Formula: C₁₁H₁₀O₂
MW: 174.1959 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00029345
CAS Number: 18515-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR