3-Fluoro-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Fluoro-2-methylphenyl)boronic acid, 3-Fluoro-2-methylbenzeneboronic acid

14275-25G 14275-5G
76907-506EA 453.01 USD
76907-506 76907-508
3-Fluoro-2-methylphenylboronic acid ≥97%
3-Fluoro-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₂
MW: 153.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368679
CAS Number: 163517-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR