4-(Cyclopentylcarbamoyl)-3-fluorobenzeneboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (4-(Cyclopentylcarbamoyl)-3-fluorophenyl)boronic acid, N-Cyclopentyl 4-borono-2-fluorobenzamide, 4-(Cyclopentylcarbamoyl)-3-fluorophenylboronic acid

14265-1G 14265-25G 14265-500MG 14265-5G
76907-494EA 173.99 USD
76907-494 76907-496 76907-498 76907-500
4-(Cyclopentylcarbamoyl)-3-fluorobenzeneboronic acid ≥97% research grade
4-(Cyclopentylcarbamoyl)-3-fluorobenzeneboronic acid
Formula: C₁₂H₁₅BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09475825
CAS Number: 957034-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR