3-Isoproproxy-4-methylphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

14241-1G 14241-25G 14241-500MG 14241-5G
76907-458EA 126.25 USD
76907-458 76907-460 76907-462 76907-464
3-Isoproproxy-4-methylphenylboronic acid ≥97% research grade
3-Isoproproxy-4-methylphenylboronic acid
Formula: C₁₀H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD11617254
CAS Number: 1451390-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR