4-Bromo-1-butoxy-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

14164-1G 14164-25G 14164-5G
76907-446EA 62.6 USD
76907-446 76907-448 76907-450
4-Bromo-1-butoxy-2-fluorobenzene ≥95%
4-Bromo-1-butoxy-2-fluorobenzene
Formula: C₁₀H₁₂BrFO
MW: 247.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907658
CAS Number: 54509-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR