3,6-Dibromo-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

14154-1G 14154-25G 14154-5G
76907-434EA 63.66 USD
76907-434 76907-436 76907-438
3,6-Dibromo-2-fluorobenzaldehyde ≥97%
3,6-Dibromo-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Br₂FO
Storage Temperature: Ambient
CAS Number: 870703-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR