4-((4-Methoxybenzyl)oxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

141323-1G 141323-25G 141323-500MG 141323-5G
76907-424EA 179.3 USD
76907-424 76907-426 76907-428 76907-430
4-((4-Methoxybenzyl)oxy)benzoic acid ≥97%
4-((4-Methoxybenzyl)oxy)benzoic acid
Formula: C₁₅H₁₄O₄
MW: 258.2692 g/mol
Storage Temperature: Ambient
CAS Number: 62290-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR