3-Ethyl-4-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

14109-1G 14109-500MG 14109-5G
76907-394EA 498.63 USD
76907-394 76907-396 76907-398
3-Ethyl-4-formylbenzoic acid ≥97%
3-Ethyl-4-formylbenzoic acid
Formula: C₁₀H₁₀O₃
MW: 178.185 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20660436
CAS Number: 1187028-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR