3-(4-Bromo-2,6-dimethylphenylcarbamoyl)-4-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14086-1G 14086-25G 14086-5G
76907-382EA 64.72 USD
76907-382 76907-384 76907-386
3-(4-Bromo-2,6-dimethylphenylcarbamoyl)-4-fluorophenylboronic acid ≥97%
3-(4-Bromo-2,6-dimethylphenylcarbamoyl)-4-fluorophenylboronic acid
Formula: C₁₅H₁₄BBrFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11856002
CAS Number: 1451391-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR