3-Acetyl-5-chlorothiophene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(5-Chlorothiophen-3-yl)ethanone

14065-25G 14065-5G
76907-368EA 442.4 USD
76907-368 76907-370
3-Acetyl-5-chlorothiophene ≥97%
3-Acetyl-5-chlorothiophene

Precaución: for Research Use Only

Formula: C₆H₅ClOS
Storage Temperature: Refrigerator
MDL Number: MFCD00082792
CAS Number: 58119-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR