4-(4-Bromophenoxy)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

14032-1G 14032-25G 14032-500MG 14032-5G
76907-348EA 290.69 USD
76907-348 76907-350 76907-352 76907-354
4-(4-Bromophenoxy)phenylboronic acid ≥97% research grade
4-(4-Bromophenoxy)phenylboronic acid
Formula: C₁₂H₁₀BBrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD23713087
CAS Number: 1451393-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR