3-tert-Butylcarbamoyl-4-fluorobenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Tert-Butylcarbamoyl)-4-fluorophenylboronic acid

14024-1G 14024-25G 14024-5G
76907-336EA 62.6 USD
76907-336 76907-338 76907-340
3-tert-Butylcarbamoyl-4-fluorobenzeneboronic acid ≥97%
3-tert-Butylcarbamoyl-4-fluorobenzeneboronic acid
Formula: C₁₁H₁₅BFNO₃
MW: 239.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235045
CAS Number: 874219-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR