3-Benzylcarbamoyl-4-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Benzylcarbamoyl)-4-fluorophenylboronic acid

14019-1G 14019-25G 14019-5G
76907-324EA 115.64 USD
76907-324 76907-326 76907-328
3-Benzylcarbamoyl-4-fluorophenylboronic acid ≥97%
3-Benzylcarbamoyl-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD08235041
CAS Number: 874219-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR