3-Carbamoyl-4-fluorobenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Aminocarbonyl)-4-fluorophenylboronic acid, (3-Carbamoyl-4-fluorophenyl)boronic acid

14012-1G 14012-25G 14012-5G
76907-312EA 114.58 USD
76907-312 76907-314 76907-316
3-Carbamoyl-4-fluorobenzeneboronic acid ≥97%
3-Carbamoyl-4-fluorobenzeneboronic acid
Formula: C₇H₇BFNO₃
MW: 182.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235053
CAS Number: 874219-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR