4-Bromo-2-chloro-3-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

14000-1G 14000-25G 14000-5G
76907-300EA 98.67 USD
76907-300 76907-302 76907-304
4-Bromo-2-chloro-3-iodopyridine ≥97%
4-Bromo-2-chloro-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClIN
Storage Temperature: Refrigerator
MDL Number: MFCD08060939
CAS Number: 916203-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR