5-Cyano-3-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile, 3-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Cyanopyridine-5-boronic acid Pinacol ester, 3-Cyanopyridine-5-boronic acid pinacol ester, 2-(5-Cyano-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

13772-1G 13772-25G 13772-5G
76907-280EA 67.9 USD
76907-280 76907-282 76907-284
5-Cyano-3-pyridineboronic acid pinacol ester ≥97%
5-Cyano-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD07780755
CAS Number: 402718-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR