Benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
13726-1G
13726-25G
13726-5G
76907-266EA
74.27
USD
76907-266
76907-268
76907-270
Benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%
Benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Formula:
C₂₀H₂₃BO₄ Storage Temperature: Ambient |
MDL Number:
MFCD06795667 CAS Number: 934984-01-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...