4-Benzyloxy-3-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Benzyloxy-3-methoxybenzeneboronic acid, (4-(Benzyloxy)-3-methoxyphenyl)boronic acid

13627-25G 13627-5G
76907-260EA 251.44 USD
76907-260 76907-262
4-Benzyloxy-3-methoxyphenylboronic acid ≥97%
4-Benzyloxy-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅BO₄
MW: 258.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09027252
CAS Number: 243990-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR