3-Fluoro-4-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Fluoro-4-(trifluoromethyl)phenyl)boronic acid, 3-Fluoro-4-trifluoromethylphenylboronic acid

13583-25G 13583-5G
76907-224EA 346.92 USD
76907-224 76907-226
3-Fluoro-4-(trifluoromethyl)phenylboronic acid ≥97%
3-Fluoro-4-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₄O₂
MW: 207.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08706261
CAS Number: 864759-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR