4-(3-Fluorophenylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(3-Fluorophenylcarbamoyl)benzeneboronic acid, (4-((3-Fluorophenyl)carbamoyl)phenyl)boronic acid

13482-25G 13482-5G
76907-192EA 605.78 USD
76907-192 76907-194
4-(3-Fluorophenylcarbamoyl)phenylboronic acid ≥97%
4-(3-Fluorophenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BFNO₃
MW: 259.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027204
CAS Number: 874288-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR